Assay Details
Target Gene Details
Entrez Gene ID: | 11666 |
Gene Name: | ATP-binding cassette, sub-family D (ALD), member 1 |
Gene Aliases: |
ALDP, Ald, Aldgh |
Location: |
Chr.X:73716597-73738287 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abcd1 | NM_007435.1 | 10 | 3077 | NP_031461.1 |
| AF213385.1 | 1 | 307 | AAF31425.1 | |
| AK009552.1 | 1 | 603 | ||
| AK088792.1 | 10 | 2947 | BAC40574.1 | |
| AK179768.1 | 1 | 234 | ||
| BC011273.1 | 10 | 2896 | AAH11273.1 | |
| BC079840.1 | 10 | 2955 | AAH79840.1 | |
| W89869.1 | 1 | 307 | ||
| Z33637.1 | 10 | 3077 | CAA83917.1 |
More Information
Set Membership: |
|

Genomic Map