Assay Details
Target Gene Details
Entrez Gene ID: | 11551 |
Gene Name: | adrenergic receptor, alpha 2a |
Gene Aliases: |
AW122659, Adra-2, Adra-2a, alpha(2A)AR, alpha2-C10, alpha2A, alpha2A-AR |
Location: |
Chr.19:54045182-54048982 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adra2a | NM_007417.4 | 1 | 2528 | NP_031443.4 |
| AK046802.1 | 2 | 2322 | ||
| AK138518.1 | 1 | 42 | ||
| AK140461.1 | 2 | 2322 | ||
| AK141573.1 | 1 | 2478 | BAE24742.1 | |
| BC138531.1 | 1 | 2144 |
More Information
Set Membership: |
|

Genomic Map