Assay Details
Target Gene Details
Entrez Gene ID: | 11554 |
Gene Name: | adrenergic receptor, beta 1 |
Gene Aliases: |
Adrb-1, beta-AR |
Location: |
Chr.19:56722372-56724862 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adrb1 | NM_007419.2 | 1 | 1659 | NP_031445.2 |
| AK018378.1 | 1 | 1477 | BAB31185.1 |
More Information
Set Membership: |
|

Genomic Map