Assay Details
Target Gene Details
Entrez Gene ID: | 12803 |
Gene Name: | ciliary neurotrophic factor |
Gene Aliases: |
AI429687 |
Location: |
Chr.19:12763528-12765632 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cntf | NM_170786.2 | 1 | 235 | NP_740756.1 |
| AK041204.1 | 1 | 70 | BAC30862.1 | |
| BC027539.1 | 1 | 235 | AAH27539.1 | |
| W08735.1 | 1 | 187 |
Target Gene Details
Entrez Gene ID: | 664779 |
Gene Name: | Zfp91-Cntf readthrough transcript (NMD candidate) |
Gene Aliases: |
Zfp91-Cntf |
Location: |
Chr.19:12763528-12796123 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp91Cntf | NR_024093.1 |

Genomic Map