Assay Details
Target Gene Details
Entrez Gene ID: | 23789 |
Gene Name: | coronin, actin binding protein 1B |
Gene Aliases: |
- |
Location: |
Chr.19:4148612-4154035 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Coro1b | NM_011778.2 | 11 | 1550 | NP_035908.1 |
| AF143956.1 | 11 | 1557 | AAD32704.1 | |
| AK008947.1 | 7 | 990 | BAB25985.1 | |
| AK149639.1 | 11 | 1560 | BAE29000.1 | |
| AK159329.1 | 9 | 1242 | ||
| AK185002.1 | ||||
| AK213696.1 | ||||
| BC131944.1 | 11 | 1497 | ||
| BC132617.1 | 11 | 1497 |
More Information
Set Membership: |
|

Genomic Map