Assay Details
Target Gene Details
Entrez Gene ID: | 18426 |
Gene Name: | OVO homolog-like 1 (Drosophila) |
Gene Aliases: |
BB147136, Ovo1, movo1 |
Location: |
Chr.19:5549137-5560575 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ovol1 | NM_019935.3 | 4 | 1144 | NP_064319.1 |
| AF134804.1 | 4 | 1160 | AAD29689.1 | |
| AF134805.1 | 4 | 1126 | AAD29690.1 | |
| AK003801.1 | 4 | 1179 | BAB23003.1 | |
| AK028577.1 | 4 | 1145 | BAC26015.1 | |
| AK132478.1 | 4 | 1145 | BAE21188.1 | |
| AK132505.1 | 4 | 1183 | BAE21208.1 | |
| AK132506.1 | 4 | 1179 | BAE21209.1 | |
| AK144952.1 | 4 | 1179 | BAE26151.1 | |
| BC021411.1 | 4 | 1224 | AAH21411.1 |

Genomic Map