Assay Details
Target Gene Details
Entrez Gene ID: | 245631 |
Gene Name: | melanoma associated antigen (mutated) 1-like 1 |
Gene Aliases: |
9430086K21Rik |
Location: |
Chr.X:139210030-139238335 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mum1l1 | NM_001164630.1 | 7 | 2355 | NP_001158102.1 |
| NM_001164631.1 | 4 | 1937 | NP_001158103.1 | |
| NM_001164632.1 | 5 | 2098 | NP_001158104.1 | |
| NM_001164633.1 | 5 | 2037 | NP_001158105.1 | |
| NM_175541.3 | 6 | 2278 | NP_780750.2 | |
| AK035086.1 | 1 | 1106 | BAC28940.1 | |
| BC053077.1 | 1 | 245 | AAH53077.1 | |
| BC096534.1 | 1 | 1280 | AAH96534.1 | |
| BC120584.1 | 1 | 1201 | ||
| BC137665.1 | 1 | 1201 |
More Information
Set Membership: |
|

Genomic Map