Assay Details
Target Gene Details
Entrez Gene ID: | 67790 |
Gene Name: | RAB39B, member RAS oncogene family |
Gene Aliases: |
6330580M05Rik |
Location: |
Chr.X:75572045-75578231 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab39b | NM_175122.6 | 2 | 1432 | NP_780331.1 |
| AK039013.1 | 2 | 1406 | BAC30206.1 | |
| AK048981.1 | 2 | 1406 | BAC33503.1 | |
| AK160737.1 | 2 | 1406 | BAE35978.1 | |
| BC050853.1 | 2 | 1426 | AAH50853.1 | |
| BC052472.1 | 2 | 1357 | AAH52472.1 |

Genomic Map