Assay Details
Target Gene Details
Entrez Gene ID: | 319931 |
Gene Name: | RIKEN cDNA A330032B11 gene |
Gene Aliases: |
- |
Location: |
Chr.19:37173843-37196541 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| A330032B11Rik | NR_045329.1 | |||
| AK039236.1 | ||||
| AK039361.1 |
Target Gene Details
Entrez Gene ID: | 208922 |
Gene Name: | cytoplasmic polyadenylation element binding protein 3 |
Gene Aliases: |
4831444O18Rik, CPE-BP3, mKIAA0940 |
Location: |
Chr.19:37019341-37207619 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cpeb3 | NM_001290826.1 | 2 | 703 | NP_001277755.1 |
| NM_001290827.1 | 2 | 352 | NP_001277756.1 | |
| NM_001290828.1 | 2 | 414 | NP_001277757.1 | |
| NM_001290829.1 | 2 | 295 | NP_001277758.1 | |
| NM_198300.3 | 2 | 414 | NP_938042.2 | |
| AB093274.1 | 2 | 343 | BAC41458.1 | |
| AK029261.1 | 2 | 296 | ||
| AK044639.1 | 2 | 416 | ||
| AK147243.1 | 2 | 704 | BAE27791.1 | |
| AK161513.1 | 2 | 390 | BAE36436.1 | |
| AY313774.1 | 2 | 236 | AAQ20843.1 | |
| BB642665.1 | 2 | 416 | ||
| BC128377.1 |
More Information
Set Membership: |
|

Genomic Map