Assay Details
Target Gene Details
Entrez Gene ID: | 100042480 |
Gene Name: | NHS-like 2 |
Gene Aliases: |
1110062M06Rik, 6430511F03, AI225852, AW488865, EG621083, ENSMUSG00000073037, Gm10456 |
Location: |
Chr.X:101848669-102092055 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nhsl2 | NM_001163610.1 | NP_001157082.1 | ||
| AK158124.1 | BAE34370.1 |
Target Gene Details
Entrez Gene ID: | 331474 |
Gene Name: | retrotransposon gag domain containing 4 |
Gene Aliases: |
6430402L03Rik, Mar5, Mart5, mKIAA2001 |
Location: |
Chr.X:102066542-102071307 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rgag4 | NM_001278534.1 | 1 | 3177 | NP_001265463.1 |
| AK032151.1 | 1 | 3067 | BAC27726.1 | |
| AK220436.1 | 1 | 3174 | BAD90267.1 | |
| BC132391.1 | 1 | 3031 | ||
| BC137795.1 | 1 | 3031 |
More Information
Set Membership: |
|

Genomic Map