Assay Details
Target Gene Details
Entrez Gene ID: | 20964 |
Gene Name: | synapsin I |
Gene Aliases: |
Syn-1, Syn1-S |
Location: |
Chr.X:20860511-20920918 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Syn1 | NM_001110780.1 | NP_001104250.1 | ||
| NM_013680.4 | NP_038708.3 | |||
| BC022954.1 | AAH22954.1 |
Target Gene Details
Entrez Gene ID: | 21857 |
Gene Name: | tissue inhibitor of metalloproteinase 1 |
Gene Aliases: |
Clgi, EPA, TIMP-1, TPA-S1, Timp |
Location: |
Chr.X:20870166-20874737 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Timp1 | NM_001044384.1 | 6 | 719 | NP_001037849.1 |
| NM_001294280.2 | 5 | 639 | NP_001281209.1 | |
| NM_011593.2 | 6 | 716 | NP_035723.2 | |
| AA184230.1 | 4 | 485 | ||
| AY622853.1 | 3 | 253 | AAT46408.1 | |
| BC008107.1 | 6 | 621 | AAH08107.1 | |
| BC034260.1 | 6 | 621 | AAH34260.1 | |
| BC051260.1 | 6 | 601 | AAH51260.1 | |
| BU756519.1 | 1 | 187 | ||
| M17243.1 | 6 | 582 | AAA40471.1 | |
| V00755.1 | 1 | 53 | CAA24132.1 | |
| X04684.1 | 6 | 658 | CAA28386.1 |
More Information
Set Membership: |
|

Genomic Map