Assay Details
Target Gene Details
Entrez Gene ID: | 66514 |
Gene Name: | asparaginase like 1 |
Gene Aliases: |
2410004D18Rik, ALP, ALP1, AU040643, AW060375 |
Location: |
Chr.19:9111719-9135566 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asrgl1 | NM_025610.3 | 7 | 1785 | NP_079886.2 |
| AK030209.1 | 7 | 1786 | BAC26844.1 | |
| BC016106.1 | 5 | 1537 | AAH16106.1 |
More Information
Set Membership: |
|

Genomic Map