Assay Details
Target Gene Details
Entrez Gene ID: | 67804 |
Gene Name: | sorting nexin 2 |
Gene Aliases: |
0610030A03Rik |
Location: |
Chr.18:53176287-53220860 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Snx2 | NM_026386.1 | 15 | 1764 | NP_080662.1 |
| AK002692.1 | 15 | 1764 | BAB22287.1 | |
| AK010572.1 | 15 | 1761 | BAB27035.1 | |
| AK075929.1 | 15 | 1761 | BAC36060.1 | |
| AK145840.1 | 15 | 1815 | BAE26688.1 | |
| AK153525.1 | 15 | 1761 | BAE32065.1 |
More Information
Set Membership: |
|

Genomic Map