Assay Details
Target Gene Details
Entrez Gene ID: | 67116 |
Gene Name: | CUE domain containing 2 |
Gene Aliases: |
3010002G01Rik |
Location: |
Chr.19:46329812-46338660 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cuedc2 | NM_001164290.1 | 10 | 1024 | NP_001157762.1 |
| NM_001164291.1 | 9 | 916 | NP_001157763.1 | |
| NM_001164292.1 | 9 | 913 | NP_001157764.1 | |
| NM_001164293.1 | 9 | 883 | NP_001157765.1 | |
| NM_001164294.1 | 9 | 862 | NP_001157766.1 | |
| NM_001164295.1 | 9 | 859 | NP_001157767.1 | |
| NM_024192.2 | 10 | 1015 | NP_077154.2 | |
| AB232359.1 | 8 | 814 | BAE19943.1 | |
| AK003457.1 | 9 | 818 | ||
| AK008842.1 | BAB25923.3 | |||
| AK013884.1 | 9 | 874 | BAB29035.1 | |
| BC003442.1 | 10 | 979 | AAH03442.1 |

Genomic Map