Assay Details
Target Gene Details
Entrez Gene ID: | 13082 |
Gene Name: | cytochrome P450, family 26, subfamily a, polypeptide 1 |
Gene Aliases: |
Cyp26, P450RA, P450RAI, RAH |
Location: |
Chr.19:37697808-37701536 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp26a1 | NM_007811.2 | 7 | 1509 | NP_031837.2 |
| AA239785.1 | ||||
| AF115769.1 | 7 | 1433 | AAD17217.1 | |
| BC012673.1 | 7 | 1469 | AAH12673.1 | |
| CJ217321.1 | 1 | 126 | ||
| Y12657.1 | 7 | 1463 | CAA73206.1 |
More Information
Set Membership: |
|

Genomic Map