Assay Details
Target Gene Details
Entrez Gene ID: | 13712 |
Gene Name: | ELK1, member of ETS oncogene family |
Gene Aliases: |
Elk-1 |
Location: |
Chr.X:20933395-20950608 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elk1 | NM_007922.5 | 6 | 2113 | NP_031948.4 |
| AK132354.1 | 6 | 2106 | BAE21121.1 |
More Information
Set Membership: |
|

Genomic Map