Assay Details
Target Gene Details
Entrez Gene ID: | 58242 |
Gene Name: | nudix (nucleoside diphosphate linked moiety X)-type motif 11 |
Gene Aliases: |
DIPP3, DIPP3b |
Location: |
Chr.X:6047479-6054751 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nudt11 | NM_021431.2 | 2 | 1178 | NP_067406.2 |
| AB041576.1 | 2 | 924 | BAA95060.1 | |
| AK036077.1 | 2 | 509 | ||
| AK048263.1 | 2 | 1181 | ||
| AK053310.1 | 2 | 958 | BAC35339.1 | |
| AY152853.1 | 2 | 924 | AAN41645.1 | |
| BC071274.1 | 2 | 982 | AAH71274.1 |
More Information
Set Membership: |
|

Genomic Map