Assay Details
Target Gene Details
Entrez Gene ID: | 24068 |
Gene Name: | steroid receptor RNA activator 1 |
Gene Aliases: |
AA959952, Sra, Srap, Straa1, Strra1 |
Location: |
Chr.18:36667187-36670311 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sra1 | NM_001164406.1 | 5 | 603 | NP_001157878.1 |
| NM_025291.3 | 5 | 706 | NP_079567.2 | |
| NR_028355.1 | 4 | 824 | ||
| AF092039.1 | 4 | 610 | ||
| AK007302.1 | 5 | 692 | BAB24943.1 | |
| AK010375.1 | 5 | 692 | BAB26893.1 | |
| AK154232.1 | 5 | 676 | BAE32451.1 | |
| AK180031.1 | 2 | 229 | ||
| BC026480.1 | 3 | 485 | AAH26480.1 | |
| BC048362.1 | 5 | 678 | AAH48362.1 | |
| BQ748094.1 | 3 | 374 |

Genomic Map