Assay Details
Target Gene Details
Entrez Gene ID: | 18648 |
Gene Name: | phosphoglycerate mutase 1 |
Gene Aliases: |
2310050F24Rik, Pgam-1 |
Location: |
Chr.19:41911871-41918665 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pgam1 | NM_023418.2 | 1 | 53 | NP_075907.2 |
| AF283667.1 | AAG13955.1 | |||
| BC002241.1 | AAH02241.1 | |||
| BC005661.1 | AAH05661.1 | |||
| BC106139.1 | AAI06140.1 |
More Information
Set Membership: |
|

Genomic Map