Assay Details
Target Gene Details
Entrez Gene ID: | 16431 |
Gene Name: | integral membrane protein 2A |
Gene Aliases: |
AW108051, Bricd2a, E25, Itm2 |
Location: |
Chr.X:107397195-107403360 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itm2a | NM_008409.2 | 6 | 1242 | NP_032435.2 |
| AK014024.1 | 6 | 1010 | BAB29119.1 | |
| AK051956.1 | 6 | 1221 | BAC34819.1 | |
| AK135229.1 | 6 | 1194 | ||
| AK167351.1 | 6 | 1243 | BAE39450.1 | |
| BC031420.1 | 6 | 1209 | AAH31420.1 | |
| L38971.1 | 6 | 1245 | AAC37700.1 |

Genomic Map