Assay Details
Target Gene Details
Entrez Gene ID: | 66240 |
Gene Name: | potassium voltage-gated channel, Isk-related family, member 1-like, pseudogene |
Gene Aliases: |
1500015C14Rik, Kcne5, Mink |
Location: |
Chr.X:142304753-142306198 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcne1l | NM_021487.1 | 1 | 729 | NP_067462.1 |
| AJ131398.1 | 1 | 729 | CAB58362.1 | |
| AK005246.1 | 1 | 712 | ||
| AK014537.1 | 1 | 564 | BAC25441.1 | |
| BC028942.1 | 1 | 707 | AAH28942.1 |
More Information
Set Membership: |
|

Genomic Map