Assay Details
Target Gene Details
Entrez Gene ID: | 11632 |
Gene Name: | aryl-hydrocarbon receptor-interacting protein |
Gene Aliases: |
AA408703, AW476050, Ara9, D19Bwg1412e, Fkbp16, Xap2 |
Location: |
Chr.19:4113756-4127236 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aip | NM_001276284.1 | 6 | 1921 | NP_001263213.1 |
| NM_016666.3 | 7 | 2020 | NP_057875.1 |
Target Gene Details
Entrez Gene ID: | 18739 |
Gene Name: | phosphatidylinositol transfer protein, membrane-associated 1 |
Gene Aliases: |
DRES9, Mpt-1, Nir-2, PITPnm 1, Pitpnm, R75447, Rd9, RdgB, RdgB1 |
Location: |
Chr.19:4099998-4113965 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pitpnm1 | NM_008851.4 | 23 | 4351 | NP_032877.1 |
| NR_075078.1 | 24 | 4045 | ||
| AF006467.1 | 24 | 3926 | AAB84393.1 | |
| AK155377.1 | 23 | 4824 | ||
| AK183265.1 | 1 | 188 | ||
| AK189965.1 | 1 | 188 | ||
| AK200663.1 | 1 | 188 | ||
| AK209641.1 | 1 | 188 | ||
| AK217085.1 | 1 | 188 | ||
| AK218358.1 | 1 | 188 | ||
| BC044893.1 | 23 | 3856 | AAH44893.1 | |
| BC048150.1 | 23 | 4005 | AAH48150.1 | |
| X98655.1 | 1 | 242 | ||
| Y08922.1 | 23 | 4352 | CAA70127.1 |
More Information
Set Membership: |
|

Genomic Map