Assay Details
Target Gene Details
Entrez Gene ID: | 226026 |
Gene Name: | structural maintenance of chromosomes 5 |
Gene Aliases: |
MLZ-453, SMC-5, Smc5l1, mKIAA0594, mSMC5 |
Location: |
Chr.19:23206441-23273908 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smc5 | NM_001252684.1 | 9 | 1264 | NP_001239613.1 |
| NM_001252685.1 | 9 | 1093 | NP_001239614.1 | |
| NM_153808.2 | 9 | 1264 | NP_722503.1 | |
| AJ534941.1 | 9 | 1185 | CAD59184.1 | |
| AK122321.1 | 2 | 210 | BAC65603.3 | |
| AK143818.1 | 9 | 1241 | ||
| AK161254.1 | 9 | 1094 | ||
| BC038345.1 | 9 | 1244 | AAH38345.1 | |
| CN530965.1 |

Genomic Map