Assay Details
Target Gene Details
Entrez Gene ID: | 70864 |
Gene Name: | family with sequence similarity 47, member C |
Gene Aliases: |
4921509A18Rik, Fam47a, Fam47a-ps |
Location: |
Chr.X:78737763-78739410 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam47c | NM_001164739.1 | 1 | 267 | NP_001158211.1 |
| AK014848.1 | 1 | 269 | ||
| BC115971.1 | 1 | 238 |

Genomic Map