Assay Details
Target Gene Details
Entrez Gene ID: | 56501 |
Gene Name: | E74-like factor 4 (ets domain transcription factor) |
Gene Aliases: |
AV314029, BC042423, ENSMUSG00000053512, Gm9907, Mef |
Location: |
Chr.X:48411049-48463132 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elf4 | NM_019680.2 | 8 | 1384 | NP_062654.2 |
| AF016714.1 | 7 | 1325 | AAD01601.1 | |
| AK155203.1 | 8 | 1625 | BAE33116.1 | |
| AK155709.1 | 8 | 1385 | BAE33395.1 | |
| BC109160.1 | 7 | 1316 | AAI09161.1 | |
| BC109171.1 | 7 | 1316 | AAI09172.1 |
More Information
Set Membership: |
|

Genomic Map