Assay Details
Target Gene Details
Entrez Gene ID: | 56501 |
Gene Name: | E74-like factor 4 (ets domain transcription factor) |
Gene Aliases: |
AV314029, BC042423, ENSMUSG00000053512, Gm9907, Mef |
Location: |
Chr.X:48411049-48463132 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elf4 | NM_019680.2 | 9 | 2912 | NP_062654.2 |
| AF016714.1 | 8 | 2853 | AAD01601.1 |
More Information
Set Membership: |
|

Genomic Map