Assay Details
Target Gene Details
Entrez Gene ID: | 194854 |
Gene Name: | predicted gene 9 |
Gene Aliases: |
- |
Location: |
Chr.X:37208502-37211041 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm9 | NM_001033234.2 | 4 | 816 | NP_001028406.1 |
| AK131663.1 | BAE20750.1 | |||
| AK145837.1 | 4 | 817 | BAE26686.1 |
More Information
Set Membership: |
|

Genomic Map