Assay Details
Target Gene Details
Entrez Gene ID: | 21912 |
Gene Name: | tetraspanin 7 |
Gene Aliases: |
1200014P11Rik, A15, AI323365, Cd231, Mxs1, PE31, R74651, TALLA, Tm4sf2 |
Location: |
Chr.X:10485116-10596604 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tspan7 | NM_019634.2 | 8 | 965 | NP_062608.2 |
| AK004776.1 | 8 | 925 | BAB23554.1 | |
| AK147306.1 | 7 | 5986 | ||
| AK147404.1 | 6 | 6263 | BAE27892.1 | |
| BC026516.1 | 7 | 764 | AAH26516.1 | |
| BC058603.1 | 8 | 921 | AAH58603.1 | |
| D26483.1 | 8 | 913 | BAA05493.1 |

Genomic Map