Assay Details
Target Gene Details
Entrez Gene ID: | 381201 |
Gene Name: | adaptor-related protein complex 5, beta 1 subunit |
Gene Aliases: |
Gm962 |
Location: |
Chr.19:5560250-5571262 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ap5b1 | NM_001033448.2 | 2 | 1808 | NP_001028620.1 |
| AK144121.1 | 2 | 1811 | BAE25712.1 | |
| AK171710.1 | 3 | 1504 | BAE42624.1 | |
| BC147482.1 | 2 | 1731 |

Genomic Map