Assay Details
Target Gene Details
Entrez Gene ID: | 64380 |
Gene Name: | membrane-spanning 4-domains, subfamily A, member 4C |
Gene Aliases: |
5830413L19Rik, AI449185, Ms4a9 |
Location: |
Chr.19:11404717-11427246 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ms4a4c | NM_029499.3 | 7 | 1095 | NP_083775.1 |
| AB026045.1 | 7 | 1080 | BAB18757.1 | |
| AF237914.2 | 7 | 1069 | AAK37596.2 | |
| AK017928.1 | 7 | 1075 | BAB31007.1 | |
| AK151045.1 | BAE30062.1 | |||
| AK172650.1 | 7 | 1067 | BAE43112.1 | |
| BC027496.1 | AAH27496.1 |
More Information
Set Membership: |
|

Genomic Map