Assay Details
Target Gene Details
Entrez Gene ID: | 67821 |
Gene Name: | ATPase, (Na+)/K+ transporting, beta 4 polypeptide |
Gene Aliases: |
1110020B02Rik |
Location: |
Chr.X:38316122-38336784 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp1b4 | NM_001290389.1 | 2 | 423 | NP_001277318.1 |
| NM_133690.3 | 2 | 423 | NP_598451.1 | |
| AF348324.1 | 2 | 278 | AAK30148.1 | |
| AF348325.1 | 2 | 278 | AAK30149.1 | |
| AK053500.1 | 2 | 362 | BAC35406.1 | |
| BC137923.1 | 2 | 376 | ||
| BC137924.1 | 2 | 376 |
More Information
Set Membership: |
|

Genomic Map