Assay Details
Target Gene Details
Entrez Gene ID: | 226089 |
Gene Name: | RAB6A GEF complex partner 1 |
Gene Aliases: |
C030046E11Rik, C130057E09Rik, Kiaa1432 |
Location: |
Chr.19:29522282-29605921 on Build GRCm38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ric1 | NM_001081319.1 | 26 | 4559 | NP_001074788.1 |
| AK021152.1 | 1 | 103 | ||
| AK048687.1 | 6 | 1657 | ||
| AK136285.1 | BAE22914.1 | |||
| AK173171.1 | 25 | 4141 | BAD32449.1 |

Genomic Map