Assay Details
Target Gene Details
Entrez Gene ID: | 29866 |
Gene Name: | calcium binding protein 2 |
Gene Aliases: |
- |
Location: |
Chr.19:4081469-4087340 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cabp2 | NM_001160253.1 | 1 | 67 | NP_001153725.1 |
| AK007488.1 | 1 | 68 |

Genomic Map