Assay Details
Target Gene Details
Entrez Gene ID: | 546729 |
Gene Name: | calcium homeostasis modulator 1 |
Gene Aliases: |
EG546729 |
Location: |
Chr.19:47141035-47144174 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Calhm1 | NM_001081271.1 | 2 | 890 | NP_001074740.1 |

Genomic Map