Assay Details
Target Gene Details
Entrez Gene ID: | 226049 |
Gene Name: | doublesex and mab-3 related transcription factor 2 |
Gene Aliases: |
Terra |
Location: |
Chr.19:25672391-25679013 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dmrt2 | NM_145831.3 | 4 | 1454 | NP_665830.1 |
| AF539811.1 | 4 | 1454 | AAN77205.1 | |
| AK044891.1 | 4 | 1455 | BAC32130.1 | |
| AK140337.1 | 4 | 1446 | BAE24344.1 | |
| AK140347.1 | 4 | 1449 | BAE24347.1 | |
| BC027669.1 | 1 | 403 | AAH27669.1 |
More Information
Set Membership: |
|

Genomic Map