Assay Details
Target Gene Details
Entrez Gene ID: | 56390 |
Gene Name: | Sjogren's syndrome/scleroderma autoantigen 1 homolog (human) |
Gene Aliases: |
1500016H19Rik, C184L |
Location: |
Chr.19:5730306-5731732 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sssca1 | NM_020491.5 | 4 | 504 | NP_065237.3 |
| AB012865.1 | 4 | 472 | BAA87050.1 | |
| AK005265.1 | 4 | 476 | BAB23917.1 | |
| AK011957.1 | 4 | 494 | BAB27938.1 | |
| AK012592.1 | 4 | 482 | BAB28340.1 | |
| AK012704.1 | 4 | 482 | BAB28423.1 | |
| BC021593.1 | 4 | 474 | AAH21593.1 | |
| BQ033851.1 | 1 | 216 |

Genomic Map