Assay Details
Target Gene Details
Entrez Gene ID: | 78526 |
Gene Name: | RIKEN cDNA C330013F16 gene |
Gene Aliases: |
- |
Location: |
Chr.X:139240226-139357738 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C330013F16Rik | NR_045455.1 | |||
| NR_045456.1 | 4 | 582 | ||
| AK021198.1 | 4 | 581 | ||
| AK133643.1 |

Genomic Map