Assay Details
Target Gene Details
Entrez Gene ID: | 72575 |
Gene Name: | RIKEN cDNA C430049B03 gene |
Gene Aliases: |
2700063P19Rik, 9430052C07Rik, mir-322, mir-351, mir-503 |
Location: |
Chr.X:53053112-53057190 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C430049B03Rik | NR_038184.1 | |||
| NR_038185.1 | ||||
| AK012483.1 | 2 | 686 | ||
| AK021262.1 |
More Information
Set Membership: |
|

Genomic Map