Assay Details
Target Gene Details
Entrez Gene ID: | 331547 |
Gene Name: | RIKEN cDNA A230072E10 gene |
Gene Aliases: |
C130073P14, Gm15139, OTTMUSG00000019231 |
Location: |
Chr.X:151313335-151341879 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| A230072E10Rik | NR_015600.2 | |||
| AK081752.1 |
Target Gene Details
Entrez Gene ID: | 279561 |
Gene Name: | WNK lysine deficient protein kinase 3 |
Gene Aliases: |
Prkwnk3, Wnk3-ps |
Location: |
Chr.X:151168777-151320194 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wnk3 | NM_001271678.1 | 22 | 5901 | NP_001258607.1 |
| NM_001271679.1 | 22 | 5760 | NP_001258608.1 | |
| BC043119.1 | 5 | 1954 | AAH43119.1 | |
| BC060731.1 | 5 | 1954 | AAH60731.1 |
More Information
Set Membership: |
|

Genomic Map