Assay Details
Target Gene Details
Entrez Gene ID: | 170745 |
Gene Name: | X-prolyl aminopeptidase (aminopeptidase P) 2, membrane-bound |
Gene Aliases: |
9030008G12Rik, mAPP |
Location: |
Chr.X:48106913-48136982 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xpnpep2 | NM_001289729.1 | 1 | 308 | NP_001276658.1 |
| NM_133213.2 | 1 | 308 | NP_573476.2 | |
| AF367247.1 | AAK52065.1 | |||
| AF428102.1 | 1 | 159 | AAL26562.1 | |
| AK078835.1 | 1 | 115 | BAC37415.1 | |
| AK085520.1 | 1 | 105 | ||
| AK087627.1 | 1 | 115 | BAC39947.1 | |
| AK164538.1 | 1 | 136 | BAE37828.1 | |
| AK172086.1 | 1 | 309 | BAE42819.1 |
More Information
Set Membership: |
|

Genomic Map