Assay Details
Target Gene Details
Entrez Gene ID: | 76041 |
Gene Name: | coiled-coil domain containing 125 |
Gene Aliases: |
5830436D01Rik, AL023012 |
Location: |
Chr.13:100668934-100697240 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc125 | NM_001168386.2 | 10 | 2417 | NP_001161858.1 |
| NM_183115.5 | 12 | 2626 | NP_898938.4 | |
| BC085249.1 | 12 | 2344 | AAH85249.1 |
Target Gene Details
Entrez Gene ID: | 12572 |
Gene Name: | cyclin-dependent kinase 7 |
Gene Aliases: |
AI323415, AI528512, C230069N13, Cdkn7, Crk4 |
Location: |
Chr.13:100697024-100730942 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk7 | NM_009874.3 | 12 | 7461 | NP_034004.2 |
| AK164567.1 | 1 | 2536 | ||
| AW120870.1 | 1 | 152 |
More Information
Set Membership: |
|

Genomic Map