Assay Details
Target Gene Details
Entrez Gene ID: | 629873 |
Gene Name: | keratin associated protein 1-4 |
Gene Aliases: |
OTTMUSG00000004966 |
Location: |
Chr.11:99582549-99583677 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krtap1-4 | NM_001039502.2 | 1 | 745 | NP_001034591.1 |
| AK131904.1 | 1 | 746 | BAE20862.1 | |
| BC150831.1 | 1 | 719 | ||
| BC150833.1 | 1 | 719 |

Genomic Map