Assay Details
Target Gene Details
Entrez Gene ID: | 74173 |
Gene Name: | RAB10, member RAS oncogene family, opposite strand |
Gene Aliases: |
1700012B15Rik, 1810036A22Rik |
Location: |
Chr.12:3235791-3250374 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab10os | NR_015551.1 | |||
| AK005891.1 | BAB24305.1 | |||
| AK007702.1 | ||||
| AK171768.1 |
More Information
Set Membership: |
|

Genomic Map