Assay Details
Target Gene Details
Entrez Gene ID: | 72123 |
Gene Name: | coiled-coil domain containing 71 like |
Gene Aliases: |
2010109K11Rik, AV375874 |
Location: |
Chr.12:32378789-32382943 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc71l | NM_001162903.1 | 1 | 1259 | NP_001156375.1 |
| AK008367.1 | 1 | 403 | BAB25631.1 | |
| AK171109.1 | 1 | 909 | ||
| BC061478.1 | 1 | 630 | AAH61478.1 |

Genomic Map