Assay Details
Target Gene Details
Entrez Gene ID: | 69109 |
Gene Name: | family with sequence similarity 58, member B |
Gene Aliases: |
1810009O10Rik, Fam58a |
Location: |
Chr.11:78750506-78751729 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam58b | NM_197989.1 | 1 | 926 | NP_932106.1 |
| AK007413.1 | 1 | 879 | ||
| AK204309.1 | 1 | 162 | ||
| BC027022.1 | 1 | 926 | AAH27022.2 |
More Information
Set Membership: |
|

Genomic Map