Assay Details
Target Gene Details
Entrez Gene ID: | 11622 |
Gene Name: | aryl-hydrocarbon receptor |
Gene Aliases: |
Ah, Ahh, Ahre, In, bHLHe76 |
Location: |
Chr.12:35497979-35534989 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ahr | NM_013464.4 | 11 | 4361 | NP_038492.1 |
| AK014512.1 | 1 | 149 | ||
| AK153678.1 | 11 | 4362 | BAE32140.1 | |
| AK155380.1 | 11 | 4413 | BAE33231.1 | |
| AK171945.1 | 1 | 1911 | ||
| AK172436.1 | 1 | 1911 |
More Information
Set Membership: |
|

Genomic Map