Assay Details
Target Gene Details
Entrez Gene ID: | 73681 |
Gene Name: | tRNA methyltransferase 11 |
Gene Aliases: |
2410075D05Rik, 3110045I18Rik, AW213713 |
Location: |
Chr.10:30534225-30602147 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trmt11 | NM_028604.2 | 13 | 1505 | NP_082880.2 |
| AF532979.1 | 13 | 1505 | AAQ10286.1 | |
| AK010722.1 | 13 | 1447 | BAB27142.1 | |
| BC048703.1 | 11 | 1180 | AAH48703.1 |
More Information
Set Membership: |
|

Genomic Map