Assay Details
Target Gene Details
Entrez Gene ID: | 107605 |
Gene Name: | retinol dehydrogenase 1 (all trans) |
Gene Aliases: |
- |
Location: |
Chr.10:127759726-127768299 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rdh1 | NM_080436.3 | 4 | 1933 | NP_536684.2 |
| AK136574.1 | 4 | 1976 | BAE23056.1 | |
| AY028928.1 | 4 | 1887 | AAK30001.1 | |
| BC137930.1 | 4 | 1851 |
More Information
Set Membership: |
|

Genomic Map