Assay Details
Target Gene Details
Entrez Gene ID: | 76815 |
Gene Name: | calcium binding and coiled-coil domain 2 |
Gene Aliases: |
2410154J16Rik, C77254, Ndp52, Ndp52l1 |
Location: |
Chr.11:96098916-96124367 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Calcoco2 | NM_001271018.1 | 6 | 1042 | NP_001257947.1 |
| AF490344.1 | 7 | 830 | AAO84502.1 | |
| AK010816.1 | 7 | 830 | BAB27200.1 | |
| AK145542.1 | 6 | 1043 | ||
| AK166710.1 | 7 | 770 | BAE38963.1 | |
| AK166756.1 | 7 | 748 | BAE38997.1 | |
| AK166939.1 | 7 | 977 | ||
| AK167009.1 | 7 | 980 | ||
| AK167172.1 | BAE39309.1 | |||
| AK167291.1 | 7 | 770 | ||
| BY756926.1 | 1 | 160 |

Genomic Map