Assay Details
Target Gene Details
Entrez Gene ID: | 72180 |
Gene Name: | zinc finger protein 661 |
Gene Aliases: |
2810405K07Rik, A1-5, Znf2 |
Location: |
Chr.2:127574617-127587135 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp661 | NM_001111029.1 | 5 | 476 | NP_001104499.1 |
| NM_028141.3 | 6 | 693 | NP_082417.3 | |
| AK033122.1 | 5 | 478 | BAC28161.1 | |
| AK134651.1 | 6 | 1746 | ||
| BC052876.1 | 6 | 697 | AAH52876.1 |
More Information
Set Membership: |
|

Genomic Map